RETSCH PM 100 Planterary Ball Mill Grinding Jars
Specially designed for extreme working conditions such as long-term trials, wet grinding, high mechanical loads and maximum speeds as well as mechanical alloying
$8863.23 - $8863.23
Specifications
Product Type Grinding Jar
For Use With (Application) Ideal for extreme working conditions such as long-term trials, wet grinding, high mechanical loads and maximum speeds as well as mechanical alloying
For Use With (Equipment) PM 100 and PM 200 planetary ball mill
Includes Nonslip seating, built-in anti-rotation device, conical base centering, O-ring, dustproof seal
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Material Price Quantity  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Material Price Quantity  
08-418-167
View Documents
RETSCH
014620228
16.9 oz. 500 mL Stainless Steel
Each for $8,863.23
Description
Description
For use with Retsch PM 100 and PM 200 Planetary Ball Mills.
  • Safe, nonslip seating with built-in anti-rotation device and conical base centering
  • With O-ring for gastight and dustproof seal
  • Lid is easy to grip and open
Specifications
Specifications
Grinding Jar
Ideal for extreme working conditions such as long-term trials, wet grinding, high mechanical loads and maximum speeds as well as mechanical alloying
PM 100 and PM 200 planetary ball mill
Nonslip seating, built-in anti-rotation device, conical base centering, O-ring, dustproof seal
SDS
missing translation for 'documents'
missing translation for 'documents'