Corning™ PYREX™ Reusable Class A Volumetric Pipets
Borosilicate-glass with fire-polished top and tip manufactured and calibrated To Contain(TC) and To Deliver(TD)
Supplier:  Corning™ 71036
 View more versions of this product
Catalog No. 13-646-41
Explore available promotions
Alert:
Do not pipet by mouth. Use mechanical pipetting device.
Description
Description
Calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969. Each pipet is individually serialized and supplied with a Certificate of Identification and Capacity, traceable to NIST standards. These precision pipets are manufactured and calibrated “To Contain” (bottom line) and “To Deliver” (top line) to specifications for Class A volumetric ware. Sizes 1 mL through 25 mL are color-coded with colored graduations enameled onto the glass.
Do not pipet by mouth. We suggest using a mechanical pipetting device.
Tested and calibrated in an ISO/IEC 17025 certified laboratory.
Certifications
NIST traceable
Compliance
Calibrated To Contain (TC - bottom line) and To Deliver (TD - top line)
Specifications
Specifications
Autoclavable
A
Non-sterile
Round
0.2 oz.
6 mL
Volumetric Pipet
Mechanical Pipetting Device
ASTM E542, ASTM E969
Borosilicate Glass
16.53 in.
±0.01 mL
Orange
42 cm
Reusable