Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Fisherbrand™ Reusable Class A Volumetric Pipets
Available in either standard or serialized models.
$50.31 - $1076.34
Spécifications
Class | A |
---|---|
Material | Borosilicate Glass |
Graduated | Yes |
Graduation Color | Amber |
Tip Style | Tuf-Temp™ treated |
Numéro de catalogue | Numéro du manufacturier. | Accuracy | Volume (Metric) | Type | Min. Flow Time | Prix | Quantité | ||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Accuracy | Volume (Metric) | Type | Min. Flow Time | Prix | Quantité | ||
13-650-3J
|
N/A | ±0.02 mL | 8 mL | Serialized Volumetric Pipet | 10 sec. |
caisse de 6 chaque for $366.00
|
|
||
13-6503X
|
N/A | ±0.02 mL | 12 mL | Serialized Volumetric Pipet | 25 sec. |
caisse de 6 chaque for $1,076.34
|
|
||
13-650-2S
|
N/A | ±0.05 mL | 50 mL | Volumetric Pipet | 25 sec. |
chaque for $69.86
caisse de 12 chaque for $838.32
|
|
||
13-650-3B
|
N/A | ±0.006 mL | 1 mL | Serialized Volumetric Pipet | 8 sec. |
caisse de 6 chaque for $240.69
|
|
||
13-650-3C
|
N/A | ±0.006 mL | 2 mL | Serialized Volumetric Pipet | 8 sec. |
caisse de 6 chaque for $284.03
|
|
||
13-650-3V
|
N/A | ±0.08 mL | 100 mL | Serialized Volumetric Pipet | 30 sec. |
caisse de 6 chaque for $767.31
|
|
||
13-650-2L
|
N/A | ±0.02 mL | 10 mL | Volumetric Pipet | 15 sec. |
caisse de 12 chaque for $185.24
|
|
||
13-6502X
|
N/A | ±0.02 mL | 12 mL | Volumetric Pipet | 25 sec. |
caisse de 12 chaque for $760.93
|
|
||
13-650-2W
|
N/A | ±0.16 mL | 200 mL | Volumetric Pipet | 30 sec. |
chaque for $97.21
caisse de 4 chaque for $388.84
|
|
||
13-650-3S
|
N/A | ±0.05 mL | 50 mL | Serialized Volumetric Pipet | 25 sec. |
caisse de 6 chaque for $649.93
|
|
||
13-650-3F
|
N/A | ±0.01 mL | 5 mL | Serialized Volumetric Pipet | 8 sec. |
caisse de 6 chaque for $361.76
|
|
||
13-650-2R
|
N/A | ±0.05 mL | 40 mL | Volumetric Pipet | 25 sec. |
chaque for $58.62
caisse de 12 chaque for $703.44
|
|
||
13-650-2B
|
N/A | ±0.006 mL | 1 mL | Volumetric Pipet | 8 sec. |
caisse de 12 chaque for $285.00
|
|
||
13-650-3H
|
N/A | ±0.01 mL | 7 mL | Serialized Volumetric Pipet | 10 sec. |
caisse de 6 chaque for $366.00
|
|
||
13-650-3K
|
N/A | ±0.02 mL | 9 mL | Serialized Volumetric Pipet | 10 sec. |
caisse de 6 chaque for $366.00
|
|
||
13-650-2U
|
N/A | ±0.08 mL | 100 mL | Volumetric Pipet | 30 sec. |
chaque for $73.41
caisse de 12 chaque for $880.92
|
|
||
13-650-2K
|
N/A | ±0.02 mL | 9 mL | Volumetric Pipet | 10 sec. |
caisse de 12 chaque for $271.10
|
|
||
13-650-2A
|
N/A | ±0.006 mL | 0.5 mL | Volumetric Pipet | 5 sec. |
caisse de 12 chaque for $280.76
|
|
||
13-650-3W
|
Fisherbrand™
136503W |
±0.16 mL | 200 mL | Serialized Volumetric Pipet | 30 sec. |
chaque for $131.35
|
|
||
13-650-2T
|
N/A | ±0.08 mL | 75 mL | Volumetric Pipet | 30 sec. |
chaque for $68.62
caisse de 12 chaque for $823.44
|
|
||
13-650-2M
|
N/A | ±0.03 mL | 15 mL | Volumetric Pipet | 25 sec. |
chaque for $50.31
caisse de 12 chaque for $603.72
|
|
||
13-650-2D
|
N/A | ±0.01 mL | 3 mL | Volumetric Pipet | 8 sec. |
caisse de 12 chaque for $285.38
|
|
||
13-650-2E
|
N/A | ±0.01 mL | 4 mL | Volumetric Pipet | 8 sec. |
caisse de 12 chaque for $285.76
|
|
||
13-650-3R
|
N/A | ±0.05 mL | 40 mL | Serialized Volumetric Pipet | 25 sec. |
caisse de 6 chaque for $566.14
|
|
||
13-650-3L
|
N/A | ±0.02 mL | 10 mL | Serialized Volumetric Pipet | 15 sec. |
caisse de 6 chaque for $390.97
|
|
||
13-650-3T
|
N/A | ±0.08 mL | 75 mL | Serialized Volumetric Pipet | 30 sec. |
caisse de 6 chaque for $630.00
|
|
||
13-6502Y
|
N/A | ±0.006 mL | 2.5 mL | Volumetric Pipet | 8 sec. |
caisse de 6 chaque for $364.38
|
|
||
13-650-2C
|
N/A | ±0.006 mL | 2 mL | Volumetric Pipet | 8 sec. |
caisse de 12 chaque for $200.72
|
|
||
13-650-3Q
|
N/A | ±0.03 mL | 30 mL | Serialized Volumetric Pipet | 25 sec. |
caisse de 6 chaque for $657.66
|
|
||
13-650-2P
|
N/A | ±0.03 mL | 25 mL | Volumetric Pipet | 25 sec. |
chaque for $59.00
caisse de 12 chaque for $708.00
|
|
||
13-650-2N
|
N/A | ±0.03 mL | 20 mL | Volumetric Pipet | 25 sec. |
chaque for $55.17
caisse de 12 chaque for $662.04
|
|
||
13-650-2Q
|
N/A | ±0.03 mL | 30 mL | Volumetric Pipet | 25 sec. |
chaque for $52.45
caisse de 12 chaque for $629.40
|
|
||
13-650-2G
|
N/A | ±0.01 mL | 6 mL | Volumetric Pipet | 10 sec. |
caisse de 12 chaque for $240.83
|
|
||
13-650-2H
|
N/A | ±0.01 mL | 7 mL | Volumetric Pipet | 10 sec. |
caisse de 12 chaque for $270.38
|
|
||
13-650-3D
|
N/A | ±0.01 mL | 3 mL | Serialized Volumetric Pipet | 8 sec. |
caisse de 6 chaque for $325.21
|
|
||
13-650-3M
|
N/A | ±0.03 mL | 15 mL | Serialized Volumetric Pipet | 25 sec. |
caisse de 6 chaque for $588.83
|
|
||
13-650-2J
|
N/A | ±0.02 mL | 8 mL | Volumetric Pipet | 10 sec. |
caisse de 12 chaque for $248.31
|
|
||
13-650-3E
|
N/A | ±0.01 mL | 4 mL | Serialized Volumetric Pipet | 8 sec. |
caisse de 6 chaque for $342.59
|
|
||
13-6503Y
|
N/A | ±0.006 mL | 2.5 mL | Serialized Volumetric Pipet | 8 sec. |
caisse de 12 chaque for $434.59
|
|
||
13-650-3P
|
N/A | ±0.03 mL | 25 mL | Serialized Volumetric Pipet | 25 sec. |
caisse de 6 chaque for $531.21
|
|
||
13-650-2F
|
N/A | ±0.01 mL | 5 mL | Volumetric Pipet | 8 sec. |
caisse de 12 chaque for $282.28
|
|
||
13-650-3A
|
N/A | ±0.006 mL | 0.5 mL | Serialized Volumetric Pipet | 5 sec. |
caisse de 6 chaque for $327.41
|
|
||
13-650-3G
|
N/A | ±0.01 mL | 6 mL | Serialized Volumetric Pipet | 10 sec. |
caisse de 6 chaque for $366.00
|
|
||
Description
Available in either standard or serialized models.
- Borosilicate glass
- Volume accuracy marked on each pipet
- Permanent amber markings
- Tuf-Temp™–treated tempered tips reduce chipping
Order Info
All pipets are available as PersonaPet™ personalized pipets.
Spécifications
A | |
Yes | |
Tuf-Temp™ treated |
Borosilicate Glass | |
Amber | |
Reusable |