BUCHI FlashPure ID Silica Flash Chromatography Cartridges
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
Supplier: BUCHI 145146131
Includes: 20 Pieces
Description
- High resolution of separation due to high-quality silica
- Increased fraction purity due to a better separation
- Faster evaporation of smaller fraction volumes
- Reproducible performance from batch to batch
- Increased sample throughput due to high sample loading capacity
- Solvent saving: required purification yield is achieved faster
Specifications
Silica | |
20 Pc. | |
Useful for Conventional Applications; Polar Compounds |
Reveleris | |
12 g | |
200 psig |