MilliporeSigma™ Supelco™ RheBuild™ Kit
Kits include all of the tools and parts you need to repair your SupelPRO unit. The parts included are those most likely to be damaged, worn, or lost. Parts may vary, depending on valve model.
Supplier:  MilliporeSigma™ Supelco™ 55049
 View more versions of this product
Catalog No. 111004212
Explore available promotions
Description
Description
Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.
Specifications
Specifications
Liquid Chromatography
7725/7725 i/7726
Videos
SDS
Documents
Documents
Promotions
Promotions
RheBuild is a registered trademark of IDEX Health & Science, LLC