MilliporeSigma™ Supelco™ RheBuild™ Kit
Kits include all of the tools and parts you need to repair your SupelPRO unit. The parts included are those most likely to be damaged, worn, or lost. Parts may vary, depending on valve model.
Supplier: MilliporeSigma™ Supelco™ 55053
Description
Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.Specifications
Liquid Chromatography | |
9725/9725 i/9726 Injectors |
RheBuild is a registered trademark of IDEX Health & Science, LLC