Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
$100.34 - $102.84
Specifications
For Use With (Equipment) PEEK Tubing
Material PEEK
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
03-251-801
View Documents
Thermo Scientific™
6000054
Fitting Set, Standard Length
Pack of 5 Each for $102.84
36-102-3904 DFS Item
View Documents
Thermo Scientific™
6000055
Fitting Set, Short
Pack of 5 Each for $100.34
Description
Description
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
  • Slotted back-side of the ferrule is squeezed down onto the tube by the mating conical surface of the nut
  • May be used on 1/16 in. metal or plastic tubing reliably up to 5000 psi
  • Reusable ferrule and nut
Specifications
Specifications
PEEK Tubing
PEEK
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.