missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhenium foil, 0.25mm (0.01in) thick, 99.97% (metals basis), Thermo Scientific Chemicals

Form: Approx. 3.29g/25 x 25mm; Solid in various forms

Supplier:  Thermo Scientific Chemicals 010309FI

 View more versions of this product

Catalog No. AA10309FI


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver to Gray
7440-15-5
99.97% (Metals basis)
MFCD00011195
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
[Re]
186.21
CHEBI:49882
Foil
Rhenium
Silver gray foil
Re
50 x 50 mm
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium
23947
Odorless

Chemical Identifiers

7440-15-5
186.21
WUAPFZMCVAUBPE-UHFFFAOYSA-N
23947
rhenium
Re
MFCD00011195
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
CHEBI:49882
[Re]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-124-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only