missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rhenium slug, 12.7mm (0.5 in.) dia., 99.99% (metals basis), Thermo Scientific Chemicals
$637.34 - $637.34
Chemical Identifiers
CAS | 7440-15-5 |
---|---|
Molecular Formula | Re |
Molecular Weight (g/mol) | 186.21 |
MDL Number | MFCD00011195 |
InChI Key | WUAPFZMCVAUBPE-UHFFFAOYSA-N |
Synonym | rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets |
PubChem CID | 23947 |
ChEBI | CHEBI:49882 |
IUPAC Name | rhenium |
SMILES | [Re] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA45524KT
|
Thermo Scientific Chemicals
045524KT |
1 Ea. |
Each for $637.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-15-5 | |
186.21 | |
WUAPFZMCVAUBPE-UHFFFAOYSA-N | |
23947 | |
rhenium |
Re | |
MFCD00011195 | |
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets | |
CHEBI:49882 | |
[Re] |
Specifications
7440-15-5 | |
MFCD00011195 | |
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets | |
[Re] | |
186.21 | |
CHEBI:49882 | |
Odorless | |
Slug | |
(metals basis) |
Re | |
1 Ea. | |
WUAPFZMCVAUBPE-UHFFFAOYSA-N | |
rhenium | |
23947 | |
99.99% | |
7.5 ±2 grams | |
Rhenium slug, 12.7mm (0.5 in.) dia., ≈6.35mm long |
Safety and Handling
EINECSNumber : 231-124-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only