missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhenium wire, 0.5mm (0.02in) dia, 99.97% (metals basis), Thermo Scientific Chemicals

Form: Approx.4.13g/m

$126.92 - $1261.89

Chemical Identifiers

CAS 7440-15-5
Molecular Formula Re
Molecular Weight (g/mol) 186.21
MDL Number MFCD00011195
InChI Key WUAPFZMCVAUBPE-UHFFFAOYSA-N
Synonym rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
PubChem CID 23947
ChEBI CHEBI:49882
IUPAC Name rhenium
SMILES [Re]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10312BQ
View Documents
Thermo Scientific Chemicals
010312BQ
5 cm
Each for $126.92
 
AA10312BU
View Documents
Thermo Scientific Chemicals
010312BU
25 cm
Each for $430.97
 
AA10312BY
View Documents
Thermo Scientific Chemicals
010312BY
100 cm
Each for $1,261.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-15-5
186.21
WUAPFZMCVAUBPE-UHFFFAOYSA-N
23947
rhenium
Re
MFCD00011195
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
CHEBI:49882
[Re]

Specifications

Silver to Gray
Silver gray wire
Re
5 cm
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium
23947
Odorless
Rhenium
7440-15-5
99.97% (Metals basis)
MFCD00011195
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
[Re]
186.21
CHEBI:49882
Wire
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-124-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only