MilliporeSigma™ Supelco™ Rheodyne™ Model 7010 Injector
Number of ports: 6 and Number of positions: 2
Supplier: MilliporeSigma™ Supelco™ 58827
Description
The original Rheodyne HPLC injector, Model 7010 introduces 5 μL - 5 mL samples onto a column with excellent reproducibility (intended for complete loop filling only). Six clustered ports allow compact interconnections, minimizing tubing volume. The flat-face seal is easy to adjust and service. Operates at pressures to 7000 psi (490 kg/cm2). Supplied with a 20 μL loop; order fill port separately.Specifications
Liquid Chromatography |
Rheodyne is a registered trademark of IDEX Health & Science, LLC