MilliporeSigma™ Supelco™ Rheodyne™ Model 7125 Injector
Number of ports: 6 and Number of positions: 2
Supplier:  MilliporeSigma™ Supelco™ 58826
Catalog No. 111003014
Explore available promotions
Description
Description
Rheodyne™ Model 7125 injector comprises of a six-port sample injection valve in which the sample is loaded through a built-in needle port in front of the valve.
Specifications
Specifications
Liquid Chromatography
Videos
SDS
Documents
Documents
Promotions
Promotions
Rheodyne is a registered trademark of IDEX Health & Science, LLC