Learn More
Thermo Scientific Chemicals Rhodamine 6G perchlorate, 99%, pure, laser grade
CAS: 13161-28-9 | C28H31ClN2O7 | 543.013 g/mol
Supplier: Thermo Scientific Chemicals 419035000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
| Rhodamine 6G perchlorate | |
| 13161-28-9 | |
| C28H31ClN2O7 | |
| 547nm (in methanol) | |
| Authentic | |
| ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;perchloric acid | |
| 13651880 | |
| 543.01 | |
| Pure | |
| Glass bottle | |
| Complies | |
| 500 mg |
| laser grade, 99% | |
| 99% | |
| MFCD00013117 | |
| HDAFVOZRAUFNQH-UHFFFAOYSA-N | |
| CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.OCl(=O)(=O)=O | |
| 543.013 | |
| 528nm (in methanol) | |
| ≥98.5% | |
| 104000L/(mole.cm) (lambda max.) | |
| (for use as a laser dye) passes | |
| Pink/Red | |
| Crystalline Powder To Crystals |
Chemical Identifiers
| 13161-28-9 | |
| 543.013 | |
| HDAFVOZRAUFNQH-UHFFFAOYSA-N | |
| ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;perchloric acid |
| C28H31ClN2O7 | |
| MFCD00013117 | |
| 13651880 | |
| CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.OCl(=O)(=O)=O |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 236-099-4
TSCA : TSCA
RUO – Research Use Only