Learn More
Thermo Scientific Chemicals Rhodamine B
Supplier: Thermo Scientific Chemicals 132311000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1700 (at lambda min.) | |
210.0°C to 211.0°C | |
1.1 to 1.6 | |
MFCD00011931 | |
Basic Violet 10, C.I. 45170 | |
PYWVYCXTNDRMGF-UHFFFAOYSA-N | |
[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC | |
479.02 | |
CHEBI:52334 | |
10% max. (105°C) | |
Pure | |
Green | |
Crystalline Powder or Crystals |
Rhodamine B | |
81-88-9 | |
C28H31ClN2O3 | |
19, V,8, 669 | |
Solubility in water: soluble. Other solubilities: slightly soluble in hcl and naoh, soluble in alcohol | |
Authentic | |
[9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride | |
6694 | |
549 to 558nm (in 50:50 IMS:water) | |
479.01 | |
Glass bottle | |
100 g |
Chemical Identifiers
81-88-9 | |
479.02 | |
PYWVYCXTNDRMGF-UHFFFAOYSA-N | |
6694 | |
[9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride |
C28H31ClN2O3 | |
MFCD00011931 | |
Basic Violet 10, C.I. 45170 | |
CHEBI:52334 | |
[Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
Harmful if swallowed.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only