missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rhodanine-3-acetic Acid 98.0+%, TCI America™
Supplier: TCI America R005425G
Specifications
Rhodanine-3-acetic Acid | |
147°C | |
MFCD00005491 | |
rhodanine-3-acetic acid, rhodanine-n-acetic acid, n-carboxymethylrhodanine, 3-rhodanineacetic acid, 3-thiazolidineacetic acid, 4-oxo-2-thioxo, 3-carboxymethyl rhodanine, n-carboxymethyl rhodanine, 2-4-oxo-2-thioxothiazolidin-3-yl acetic acid, 4-oxo-2-thioxo-3-thiazolidineacetic acid, 4-oxo-2-thioxo-3-thiazolidinylacetic acid | |
OC(=O)CN1C(=O)CSC1=S | |
191.22 | |
191.22 | |
Crystalline Powder |
5718-83-2 | |
C5H5NO3S2 | |
25 g | |
JGRMXPSUZIYDRR-UHFFFAOYSA-N | |
2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid | |
79793 | |
≥98.0% (T) |
Chemical Identifiers
5718-83-2 | |
191.22 | |
JGRMXPSUZIYDRR-UHFFFAOYSA-N | |
79793 | |
OC(=O)CN1C(=O)CSC1=S |
C5H5NO3S2 | |
MFCD00005491 | |
rhodanine-3-acetic acid, rhodanine-n-acetic acid, n-carboxymethylrhodanine, 3-rhodanineacetic acid, 3-thiazolidineacetic acid, 4-oxo-2-thioxo, 3-carboxymethyl rhodanine, n-carboxymethyl rhodanine, 2-4-oxo-2-thioxothiazolidin-3-yl acetic acid, 4-oxo-2-thioxo-3-thiazolidineacetic acid, 4-oxo-2-thioxo-3-thiazolidinylacetic acid | |
2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid |
Safety and Handling
EINECSNumber : (5)-0223
TSCA : Yes