missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodanine Indicator, 0.02% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 662516

 View more versions of this product

Catalog No. 662516


Explore available promotions

Description

Description

Specifications

Specifications

Rhodanine Indicator
98.0,0.02
0.02% (w/v)
KIWUVOGUEXMXSV-UHFFFAOYSA-N
2-sulfanylidene-1,3-thiazolidin-4-one
1201546
Natural Poly Bottle
Golden/Tan
Liquid
141-84-4,7732-18-5
100.0,0.02
C3H3NOS2
C1C(=O)NC(=S)S1
133.183
CHEBI:8830
Aqueous Solution
500 mL

Chemical Identifiers

141-84-4
133.183
1201546
2-sulfanylidene-1,3-thiazolidin-4-one
C3H3NOS2
KIWUVOGUEXMXSV-UHFFFAOYSA-N
CHEBI:8830
C1C(=O)NC(=S)S1
Videos
SDS
Documents

Documents

Promotions

Promotions