missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ RiboLock RNase Inhibitor (40 U/μL)
RiboLock RNase Inhibitor specifically inhibits the activity of RNases A, B, and C, protecting RNA under a variety of reaction conditions.
$114.63 - $1,890.00
Specifications
Concentration | 40 U/μL |
---|---|
Compatible Buffer | Storage Buffer |
Product Type | RNase Inhibitor |
Product Line | RiboLock™ |
Description
RiboLock RNase Inhibitor inhibits the activity of RNases A , B, and C by binding them in a noncompetitive mode at a 1:1 ratio. It does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 as well as prokaryotic RNases I and H.
Highlights
- Performs under a wide range of reaction conditions
- Protects RNA from degradation at temperatures up to 55°C
Specifications
40 U/μL | |
Storage Buffer | |
RNase Inhibitor | |
RiboLock™ |
For Research Use Only. Not for use in diagnostic procedures.