missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cleaning Solution, Chromic-Sulfuric Acid Solution, Ricca Chemical
For Cleaning Laboratory Glassware
$211.03 - $559.66
Specifications
Form | Liquid |
---|---|
Sterility | Non-sterile |
Appearance | Dark Brown |
Boiling Point | ∼300°C |
Certifications/Compliance | OSHA, 29 CFR 1910.1200, TSCA |
Description
- Clear, brown liquid
- 7789-12-0 (Sodium Dichromate Dihydrate); 1333-82-0 (Chromium (VI) Trioxide); 7664-93-9 (Sulfuric Acid)
Specifications
Liquid | |
Dark Brown | |
OSHA, 29 CFR 1910.1200, TSCA | |
15°C to 30°C |
Non-sterile | |
∼300°C | |
Chromium (VI) Trioxide 0.8 to 1%, Sulfuric Acid 97 to 99 %, Water, Deionized 1 to 1.2%, Sodium Dichr |