missing translation for 'onlineSavingsMsg'
Learn More

Electrode Storage Solution, Ricca Chemical

For storing glass and combination pH electrodes.

Supplier:  Ricca Chemical Company 27951

 View more versions of this product

Catalog No. 2795-1


Only null left
Explore available promotions

Description

Description

For storing glass and combination pH electrodes.
Specifications

Specifications

33.8 oz.
pH Electrodes
Storing glass and combination pH electrodes
4 L
4000 mL
Storage Solution, Poly Natural
Natural Poly Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions