missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

pH Test Strips, Ricca Chemical

General-purpose pH test strips

Supplier:  Ricca Chemical Company RI88801

 View more versions of this product

Catalog No. 88-801


Explore available promotions

Description

Description

  • Available in four different pH ranges
  • Time-saving, single-use indicator strips with color-fixed test area on an easy-to-handle, durable plastic strip
  • Fast accurate results
  • Packages include a color comparison chart
  • 100 strips per pack

Specifications

Specifications

0 to 14
1
1 Ea.
pH Test Strip
For precise pH determination
Color chart
15°C to 30°C
Videos
SDS
Documents

Documents

Promotions

Promotions