missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
pH Test Strips, Ricca Chemical
General-purpose pH test strips
Supplier: Ricca Chemical Company RI88811
Description
- Available in four different pH ranges
- Time-saving, single-use indicator strips with color-fixed test area on an easy-to-handle, durable plastic strip
- Fast accurate results
- Packages include a color comparison chart
- 100 strips per pack
Specifications
0 to 6 | |
0.5 | |
100 Strips |
For precise pH determination | |
Color chart | |
pH Test Strip |