missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Synthetic Urine Concentrate, Ricca Chemical
General-use Reagent.
Supplier: Ricca Chemical Company 83625
Description
- Clear, colorless liquid
- Specific Gravity: Approx. 1.1
Specifications
NIST SRM 186, 191 | |
Synthetic Urine Concentrate | |
Magnesium Sulfate Heptahydrate 0.9 to 1.2%, Calcium Chloride <1%, Urea 17 to 21%, Sodium Chloride 6 to 10%, Water, Deionized | |
7.9 | |
Liquid |
Clear | |
Reagent | |
Cubitainer | |
20 L |