Learn More
Epredia™ Richard-Allan Scientific™ Amyloid Stain, Sodium Hydroxide Solution with dropper
Demonstrate amyloid with Epredia™ Richard-Allan Scientific™ Amyloid Stain.
Supplier: Epredia™ 87003
Description
- Alkaline Congo Red solution will stain amyloid orange-to-red and display apple-green birefringence when examined with polarizing microscope
Specifications
| Stain Kit | |
| 1 Kit |