Epredia™ Dehydrant Alcohol
Dehydrant Alcohol products are specifically designed for tissue processing and staining of histological and cytological specimens. Available in 70%, 80%, 95% and 100% blends for complete dehydration.
Supplier:  Epredia™ 6215
 View more versions of this product
Catalog No. 62-15
Explore available promotions
Description
Description
  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents
Specifications
Specifications
Alcohol
1.32 gal.
100% Dehydrant