Epredia™ Richard-Allan Scientific™ Elastic Stain, Van Gieson Stain Solution
Demonstrate the atrophy, thinning and loss of elastic tissue with Epredia™ Richard-Allan Scientific™ Elastic Stain.
Supplier:  Epredia™ 88007
 View more versions of this product
Catalog No. 22-110-649
Explore available promotions
Description
Description
  • Based on the Verhoeff Elastic technique
  • Also used for identifying the breaks, splitting and reduplication of elastic lamellae that occur in vascular diseases
  • Elastic fibers are stained blue-black to-black with red collagen and a yellow background
Specifications
Specifications
Alcoholic Hematoxylin Solution