Epredia™ Richard-Allan Scientific™ Gram Stain (Tissue) Kit, Crystal Violet Solution
Identify gram-positive and gram-negative bacteria in tissue sections with Epredia™ Richard-Allan Scientific™ Gram Stain (Tissue)
Supplier:  Epredia™ 87018
 View more versions of this product
Catalog No. 22-110-647
Explore available promotions
Description
Description
  • Gram-positive organisms stain blue to blue-black
  • Gram-negative organisms stain red
  • Tartrazine provides a yellow background stain
Specifications
Specifications
Gram Stain (Tissue) Kit