Learn More
Epredia™ Richard-Allan Scientific™ Modified Wright Stain Pack
Achieve optimum performance on Hema-Tek™ Slide Stainers with the Epredia™ Richard-Allan Scientific™ Stain Pack.
Supplier:  Epredia™ SH3005
 View more versions of this product
Catalog No. 22-050-269
Only null left
Explore available promotions
Description
Description
This system consists of a modified polychrome methylene blue-eosin stain (similar to Wright or Giemsa) and based on the original stain proposed by Romanowsky*.
Specifications
Specifications
5 Pk.
Product Suggestions
Product Suggestions