Epredia™ Mounting Medium
Store slides for prolonged periods of time without fading of stains with Epredia™ Mounting Medium.
$252.32 - $284.56
Catalog Number | Mfr. No. | Volume (English) | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (English) | Quantity | Price | Quantity | |||||
22-050-102
|
Epredia™
4111 |
1 pt. | 2/Cs. |
Case of 2 Each for $252.32
|
|
|||||
22-110-610
|
Epredia™
4112 |
4 oz. | 6/Cs. |
Case of 6 Each for $284.56
|
|
|||||
Description
- Rapid drying
- Toluene-based
- Optically clear
Specifications
Mounting Medium |