Epredia™ Richard-Allan Scientific™ Mucicarmine, Tartrazine Stain Solution
Identify primary tumor sites with the aid of Epredia™ Richard-Allan Scientific™ Mucicarmine
Supplier:  Epredia™ 87012
 View more versions of this product
Catalog No. 22-110-646
Explore available promotions
Description
Description
  • Aids in distinguishing mucin-negative undifferentiated squamous cell lesions from mucin-positive adenocarcinomas
  • Mucin is stained dark rose and the nuclei are black
  • Capsule of Cryptococcus (fungus) is stained deep rose
  • Tartrazine solution is used as a yellow cytoplasmic stain
Specifications
Specifications
Mucicarmine Stain Kit