missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Neg-50™ Frozen Section Medium
Yield high-quality sections down to -50°C without chatter using water-soluble Epredia™ Neg-50™ Frozen Section Medium. Comes in clear and five additional colors with all leaving no residue after staining.
$142.13 - $424.74
Catalog Number | Mfr. No. | Color | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Quantity | Price | Quantity | ||||
36-101-4314
|
Epredia™
6502 |
Clear | 4 oz. per bottle |
Case for $142.13
|
|
||||
22-050-451
|
Epredia™
6502B |
Blue | 4 oz.per bottle |
Case of 2 Each for $152.09
|
|
||||
36-101-4361
|
Epredia™
6502G |
Green | 4 oz. per bottle |
Case for $195.72
|
|
||||
22-046-512
|
Epredia™
6502O |
Orange | 4 oz. per bottle |
Pack of 2 Each for $274.15
|
|
||||
22050453
|
Epredia™
6502P |
Pink | 4 oz. per bottle |
Case of 2 Each for $148.75
|
|
||||
22050454
|
Epredia™
6502Y |
Yellow | 4 oz. per bottle |
Case of 2 Each for $222.03
|
|
||||
22-046-511
|
Epredia™
6506 |
Clear | 4 oz. per bottle |
Case of 6 Each for $350.78
|
|
||||
22050450
|
Epredia™
6506M |
Five Colors plus Clear | 4 oz. per bottle |
Case of 6 Each for $424.74
|
|
||||
Description
Yield high-quality sections down to -50°C without chatter using Epredia; Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
- Buffered to ensure proper alkalinity (pH = 8.0); leaves no residue after staining
- Specially formulated to prevent tissue separation from the slide
Ordering Information:
Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.