missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Epredia™ Richard-Allan Scientific™ Signature Series Eosin-Y 7111 Edge
Edge

Create an ideal visual contrast to Hematoxylin with the fast-acting Epredia™ Richard-Allan Scientific™ Signature Series Eosin-Y counterstain.

Supplier:  Epredia™ 7111

 View more versions of this product

Catalog No. 22-050-110


Edge
Explore available promotions

Description

Description

  • Bright, vivid cytoplasmic staining
  • Creates three distinct shades of pink
  • Unmatched color contrast between the nuclei and cytoplasm
  • Various shades of red and pink differentiate muscle, red blood cells and connective tissue
  • Strict quality assurance guarantees product consistency and longevity
  • Quantitative testing eliminates need for adjustments in dilution or time

Available in 5 liter packaging

  • Reduces plastic by 90%
  • Reduces stain spills
  • Increases stain stability
  • Saves 35% storage space versus traditional packaging
Specifications

Specifications

Eosin Stain
1 pt.
Videos
SDS
Documents

Documents

Promotions

Promotions