Learn More
Epredia™ Richard-Allan Scientific™ Three-Step Stain Kit
Obtain results on par with Wright-Giemsa with Epredia™ Richard-Allan Scientific™ Three Step Stain.
$727.61 - $1250.10
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|
36-101-4309
|
Epredia™
SH3300 |
Epredia Three Step Stain Kit |
Case for $727.61
|
|
|||||
|
22-050-275
|
Epredia™
SH3323 |
Solution B |
Each for $1,250.10
|
|
|||||
|
22-050-274
|
Epredia™
SH3313 |
Solution A |
Each for $1,209.23
|
|
|||||
Description
- Stain can be used to detect H. Pylori.
- Set contains easy-to-follow instructions
- Product is available in one-gallon containers to facilitate use on automated strainers
- Set contains three 500mL bottles consisting of Fixative Solution, Solution A(Eosin-Y), and Solution B(Azure A and Methylene Blue)