missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Rigidwall Cleanroom
Rigidwall cleanrooms combine class and functionality offering an elegant solution for most any cleanroom application. Easy to construct. Easy to clean.
$53723.18 - $80845.78
Specifications
Product Type | Cleanroom |
---|---|
Filter Type | 19 HEPA |
Autoclavable | Not Autoclavable |
Height (English) | 12 ft. |
Catalog Number | Mfr. No. | Length (English) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Price | Quantity | |||||
15-050-989
|
Fisherbrand™
FRWISO88X12 |
8 ft. |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
15-050-991
|
Fisherbrand™
FRWISO812X12 |
12 ft. |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Easy to construct, secure, and easy to clean
- With wall panels providing full visibility from floor to ceiling, these rooms provide a comfortable working environment.
- The modular nature of this system provides opportunity for expansion as your operation grows and offers the ability to relocate the cleanroom altogether should needs shift within your organization.
- Our Rigidwall cleanrooms meet ISO 4 to ISO 8 air cleanliness ratings.
Specifications
Cleanroom | |
19 HEPA | |
Not Autoclavable | |
12 ft. |