missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ RNase AWAY™ Surface Decontaminant

Eliminate RNase and DNA from laboratory surfaces. This product reduces dependency on carcinogenic DEPC treatments and saves time spent baking glassware.

$96.84 - $369.38

Specifications

pH 11
Composition Sodium Hydroxide with Water
View More Specs

Products 4
Catalog Number Mfr. No. Container Type Volume (English) Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Container Type Volume (English) Volume (Metric) Price Quantity  
21-236-21
View Documents
Thermo Scientific™
7000
Bottle 8.5 oz. 250 mL
Each for $96.84
Case of 12 Each for $976.80
 
21-402-178
View Documents
Thermo Scientific™
7002
Spray Bottle 16 oz. 473 mL
Each for $117.87
Case of 6 Each for $594.48
 
14-375-35
View Documents
Thermo Scientific™
MB7003
Bottle 33.875 oz. 1 L
Each for $184.69
Case of 5 Each for $836.90
 
14-754-34
View Documents
Thermo Scientific™
700511
Bottle 1.06 gal. 4 L
Each for $369.38
Case of 4 Each for $1,339.04
 
Description

Description

Eliminate RNase and DNA from laboratory surfaces. Thermo Scientific™ RNase AWAY™ is ideal for decontaminating apparatus, bench-tops, glassware and plasticware. It reduces dependency on carcinogenic DEPC treatments and saves time needed to bake glassware.

  • Use on pipettes, gel boxes, or RNA or DNA prep areas
  • Leaves no residue to interfere with gel polymerization or staining
  • Chemically stable and nonabrasive
  • Contains no strong acids
Specifications

Specifications

11
Sodium Hydroxide with Water
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'