missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rocuronium Bromide 98.0+%, TCI America™
Supplier: TCI America R0099100MG
Specifications
Rocuronium Bromide | |
169°C | |
MFCD00867768 | |
OYTJKRAYGYRUJK-FMCCZJBLSA-N | |
[(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-3-hydroxy-10,13-dimethyl-2-morpholin-4-yl-16-(1-prop-2-enylpyrrolidin-1-ium-1-yl)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate;hydrobromide | |
129893438 | |
≥98.0% (T) |
119302-91-9 | |
C32H54BrN2O4+ | |
100 mg | |
CC(=O)OC1C(CC2C1(CCC3C2CCC4C3(CC(C(C4)O)N5CCOCC5)C)C)[N+]6(CCCC6)CC=C.Br | |
610.698 | |
609.69 | |
Crystalline Powder |
Chemical Identifiers
119302-91-9 | |
610.698 | |
OYTJKRAYGYRUJK-FMCCZJBLSA-N | |
[(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-3-hydroxy-10,13-dimethyl-2-morpholin-4-yl-16-(1-prop-2-enylpyrrolidin-1-ium-1-yl)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate;hydrobromide |
C32H54BrN2O4+ | |
MFCD00867768 | |
129893438 | |
CC(=O)OC1C(CC2C1(CCC3C2CCC4C3(CC(C(C4)O)N5CCOCC5)C)C)[N+]6(CCCC6)CC=C.Br |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator