missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Rosin, gum, Thermo Scientific Chemicals
$121.96 - $121.96
Identifiants chimiques
CAS | 8050-09-7 |
---|---|
Molecular Formula | C20H30O2 |
Molecular Weight (g/mol) | 302.46 |
MDL Number | MFCD00132205 |
InChI Key | RSWGJHLUYNHPMX-UHFFFAOYNA-N |
Synonym | 4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
AC419175000
|
Thermo Scientific Chemicals
419175000 |
500 g | Glass bottle |
chaque for $121.96
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentIdentifiants chimiques
8050-09-7 | |
302.46 | |
RSWGJHLUYNHPMX-UHFFFAOYNA-N |
C20H30O2 | |
MFCD00132205 | |
4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid |
Spécifications
163 to 173mg KOH/g | |
75.0°C to 81.0°C | |
180°C | |
Glass bottle | |
MFCD00132205 | |
14,8266 | |
Solubility in water: 130mg/l (20°C) | |
302.46 | |
Rosin, gum |
8050-09-7 | |
Yellow | |
Authentic | |
C20H30O2 | |
500 g | |
4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid | |
RSWGJHLUYNHPMX-UHFFFAOYNA-N | |
Crystalline Chunks and Powder |
Sécurité et manipulation
GHS H Statement
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 232-475-7
missing translation for 'rtecsNumber' : VL0480000
missing translation for 'tsca' : TSCA
RUO – Research Use Only