missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rosin (Powder, with Diluent), Fisher Chemical
Supplier: Fisher Chemical R308500
Specifications
Rosin | |
100°C | |
2.0 | |
C20H30O2 | |
500 g | |
302.46 | |
Solid |
8050-09-7,1302-78-9 | |
Amber | |
Poly Bottle | |
MFCD00132205 | |
RSWGJHLUYNHPMX-UHFFFAOYNA-N | |
Negligible |