missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rosolic acid, Thermo Scientific Chemicals

$155.22 - $410.89

Chemical Identifiers

CAS 603-45-2
Molecular Formula C19H14O3
Molecular Weight (g/mol) 290.32
MDL Number MFCD00001624
InChI Key FYEHYMARPSSOBO-UHFFFAOYSA-N
Synonym aurin, rosolic acid, corallin, p-rosolic acid, aurine, spirit aurine, corallin spirit soluble, 4,4'-dihydroxyfuchsone, aurin no. 555, 4-bis 4-hydroxyphenyl methylene cyclohexa-2,5-dienone
PubChem CID 5100
ChEBI CHEBI:34544
IUPAC Name 4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2173714
View Documents
Thermo Scientific Chemicals
B2173714
25 g
Each for $155.22
 
AAB2173722
View Documents
Thermo Scientific Chemicals
B2173722
100 g
Each for $410.89
 
Description

Description

Rosolic acid is used as a pH indicator in the range from 5.0 to 6.8. It acts as an intermediate in the manufacturing dyes. It is involved in the activation of heme oxygenase and in the inhibition of DNA fragmentation. It finds application in the selective isolation of coliform bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rosolic acid is used as a pH indicator in the range from 5.0 to 6.8. It acts as an intermediate in the manufacturing dyes. It is involved in the activation of heme oxygenase and in the inhibition of DNA fragmentation. It finds application in the selective isolation of coliform bacteria.

Solubility
Soluble in alcohol, sodium hydroxide, potassium hydroxide and strong acids. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

603-45-2
290.32
FYEHYMARPSSOBO-UHFFFAOYSA-N
5100
4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
C19H14O3
MFCD00001624
aurin, rosolic acid, corallin, p-rosolic acid, aurine, spirit aurine, corallin spirit soluble, 4,4'-dihydroxyfuchsone, aurin no. 555, 4-bis 4-hydroxyphenyl methylene cyclohexa-2,5-dienone
CHEBI:34544
OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1

Specifications

603-45-2
MFCD00001624
2055205
aurin, rosolic acid, corallin, p-rosolic acid, aurine, spirit aurine, corallin spirit soluble, 4,4'-dihydroxyfuchsone, aurin no. 555, 4-bis 4-hydroxyphenyl methylene cyclohexa-2,5-dienone
FYEHYMARPSSOBO-UHFFFAOYSA-N
4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
5100
290.32
C19H14O3
25 g
14,881
Soluble in alcohol,sodium hydroxide,potassium hydroxide and strong acids. Insoluble in water.
OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1
290.32
CHEBI:34544
Rosolic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-041-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only