Rotating Platform
Versatile turntable demonstrates a variety of fascinating rotation principles.
Supplier: Customer Channels Group (Ccg) XBL760000A
Description
- Made of two 10 in. (25cm) diameter wood discs separated by a ball-bearing race
- Demonstrates gyroscopic motion principles when used with our Bicycle Wheel Gyroscope (S40965, sold separately)
- Students can observe the conservation of angular momentum
Specifications
Rotating Platform | |
Ideal for use as a swivel stool top | |
10 in. | |
25 cm |