BUCHI Rotavapor R-300 Rotary Evaporator with Controller and V-300 Pump
The R-300 meets the highest expectations in convenience and versatility. Its modular design allows extension of the Rotavapor R-300 to a fully integrated system where a central interface regulates each component.
Supplier:  BUCHI 11SR300252VP12
 View more versions of this product
Catalog No. 05-001-074
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Intuitive user interaction
  • Precise operation control for repeatability and safety
  • 1 L or 5 L flask capacity baths with cordless changes and cleaning
  • Up to 220 °C bath heating with 5 L bath
  • Ergonomic handle with manual or electronic lift
  • Choice of P+G coating for added protection of condenser and receiving flask
  • Unattended operation and mobile monitoring available
  • Modular system for control, vacuum, and chiller integration
  • Additional glass assemblies and configurations
  • Cloud services for optimized productivity and automated processes
Specifications
Specifications
Electronic Lift
100/120 V
10 to 280 rpm
220°C
1600 W
Vertical
I-300 Pro Interface
CE/CSA
50/60 Hz
24/40
5 mbar
5 L
Woulff Bottle, F-305 Recirculating Chiller
V-300 Diaphragm
100/120 V
607 x 429 x 947 mm
1100 W
Rotary Evaporator System
R-300/V-300/F-305