United Scientific™ Round Buret Clamps with Coated Jaws
These Round Buret Clamps with Coated Jaws are ideal for holding small objects.
Supplier:  United Scientific™ COBR3FR
Catalog No. S99452
Explore available promotions
Description
Description
  • Round jaws open from 0.5 to 1.75 in. (15 to 45 mm).
  • Attaches to rods up to 0.5 in. O.D. (13 mm).
  • Coated jaws can be rotated and locked at any angle.
Specifications
Specifications
Burette Clamp
Coated Jaws
Silver
Alloy Steel
Round
2-Prong Single Adjust
Product Suggestions
Product Suggestions