Thermo Scientific™ Round Test Tube Cells for Thermo Scientific Spectronic™ 20+ and 20D+ Spectrophotometers
Supplier:  Thermo Scientific™ 331781000
 View more versions of this product
Catalog No. 07-144-40
Explore available promotions
Description
Description
  • Optical class
  • Cat. No. 07-144-15 1/2 in. diameter test tube cell replacements for originals supplied with instrument Cat. No. 07-144-40 3/4 in. diameter test tube cells require 07-144-20 adapter and 07-144-24 light shield (sold separately)
  • Cat. No. 07-144-38 test tube cell is 1 in. diameter and requires 07-144-22 adapter and 07-144-24 light shield (sold separately)
  • Packaged in box of 12
Specifications
Specifications
Test Tube
19.05 mm
Thermo Scientific Spectronic™ 20 and 20D spectrophotometers
Optical Glass
150 mm