Thermo Scientific™ RT Anaero-Indicator
Mitsubishi™ RT Anaero-Indicator is a single-use disposable means of detecting an anaerobic atmosphere.
Supplier:  Thermo Scientific™ RE684002
Catalog No. R684002
Explore available promotions
Description
Description
Mitsubishi™ RT Anaero-Indicator is a single-use disposable means of detecting an anaerobic atmosphere.
  • The indicator is a pink pill that will become purple to blue when exposed to oxygen and then back to pink when an anaerobic atmosphere is obtained
  • Used with the AnaeroPack-Anaero gas generator (Part No. 681001) and the AnaeroPouch-Anaero gas generator (Part No. R682001)
Ordering Information:
This product is for use with the AneroPack-Anaero (R681001) in a AnaeroPack Jar (R685025 and R685077) or AnaeroPouch-Anaero (R682001) in a AnaeroPouch-Bag (R686001).
Specifications
Specifications
RT Anaero-Indicator
2°C to 25°C (refrigeration is not necessary)
Thermo Scientific Mitsubishi
25/Pk.