Bullard™ RT Series Tychem™ Respirator Hoods
RT hoods are available in two different Tychem materials and are held in place by the inflatable neck cuff (RT1 and RT2 models) or a sport material neck cuff (RT3 and RT4 models).
$966.22 - $1770.75
Specifications
Assigned Protection Factor (APF) | 1,000 |
---|---|
Product Type | Respirator Hood |
For Use With (Application) | Industrial environments, petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing |
For Use With (Equipment) | PA30, EVA, and/or RT Airline respiraor |
Catalog Number | Mfr. No. | Material | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Includes | Price | Quantity | ||||
36-100-3800
|
Bullard™
RT1 |
Tychem QC | Inflatable neck cuff |
Pack of 10 Each for $966.22
|
|
||||
36-100-3801
|
Bullard™
RT2 |
Tychem SL | Inflatable neck cuff |
Pack of 5 Each for $1,770.75
|
|
||||
19-321-718
|
Bullard™
RT3 |
Tychem QC | Sport neck cuff |
Case of 10 Each for $970.09
|
|
||||
19-321-719
|
Bullard™
RT4 |
Tychem SL | Sport neck cuff |
Case of 10 Each for $1,683.52
|
|
||||
Description
- Tychem™ QC (quality coated) is a lightweight, supple material coated with polyethylene to provide splash protection against many acids, bases, and other liquid chemicals
- Tychem SL (Saranex™ laminate) is coated with Saranex 23-P to provide protection from an even wider range of chemicals
- RT1/RT3 = Tychem QC
- RT2/RT4 = Tychem SL
- Panoramic view minimizes claustrophobic feeling sometimes experienced in other respirator hoods
- Sport or inflatable neck cuff design provides positive custom seal
- Three-channel air delivery system promotes curvature of hood for better fit
- Long inner and outer bibs for splash protection
- Extra-large acetate lens
- No suspension is required
- Breathing tube connects using a sleeve on the back of the hood and a clamp
Order Info
Lens cover available. (See Cat. No. 17-986-288)
Specifications
1,000 | |
Respirator Hood | |
Industrial environments, petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing | |
PA30, EVA, and/or RT Airline respiraor |