missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ RTK Pictogram Labels, Gloves
Effectively convey personal protection instructions and special precautionary information. Brady™ RTK Pictogram Labels are made from premium grade B-946 vinyl and are sized to fit standard blank write-on right-to-know labels.
Supplier: Brady™ 58556
Description
- Made from premium grade B-946 vinyl
- Sized to fit standard Blank Write-On Right-To-Know Labels
- Correspond to ANSI proposed Z.535 Standard
- Card-mounted for easy removal
Specifications
Acrylic, Permanent, Cold-temperature | |
Pressure-sensitive Vinyl (B-946) | |
1.125 x 1.125 in. | |
0.004 in. |
White, Blue | |
RTK Pictogram Labels, Gloves | |
2.85 x 2.85cm |