Eisco™ Rubber Cones
Designed for use with Buchner Funnels in filtration assemblies, these cones are made of flexible, quality neoprene rubber. Rubber Cones have superior chemical stability, and can withstand a wide temperature range.
Supplier: Eisco™ CH0421
Description
- Available as set of 7
Specifications
Blue | |
1, 2, 3, 4, 5, 6, 7 | |
Rubber |
Rubber cones | |
7 Pk. | |
Buchner Funnels |