missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Rubber Crucible Holder

This Rubber Crucible Holder is designed to support funnels, crucibles, and other equipment in vacuum-type filtration assemblies.

$524.44 - $524.44

Specifications

Certifications/Compliance ISO, GMP
Color Gray
Material Neoprene
For Use With (Application) Support funnels, crucibles, and other equipment in vacuum-type filtration assemblies.
Autoclavable Autoclavable
View More Specs

Products 1
Catalog Number Mfr. No. Size Diameter (Metric) Outer Diameter (Metric) Base Price Quantity  
Catalog Number Mfr. No. Size Diameter (Metric) Outer Diameter (Metric) Base Price Quantity  
01-189-173
View Documents
Fisherbrand™
10401294
Size 3 37 mm 22 mm
Case of 72 Each for $524.44
 
Description

Description

  • Made from neoprene rubber
  • Useful in filtration assemblies for supporting funnels, Gooch crucibles, and filter tubes
  • Adapter does not need to be fitted closely around the funnel stem to be functional
  • Eliminates boring of special size holes in rubber stoppers
  • Designed to fit Buchner and fritted-glass funnels, porcelain filters without headpieces, and for reverse flow cleaning of filter crucibles
  • Can be used singly or nesting with adjacent sizes
  • Autoclavable
  • Available individual sizes or an assorted set
Specifications

Specifications

ISO, GMP
Neoprene
Autoclavable
Gray
Support funnels, crucibles, and other equipment in vacuum-type filtration assemblies.
Tapered
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'