missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rubbermaid™ BRUTE™ Recycling Containers
Easy-to-maneuver
Supplier: Rubbermaid™ 353673BLUE
Description
- Contains 30% Post Consumer Recycled Resin (PCR) exceeding EPA guidelines.
- Can serve as central collection containers or shipping containers, reducing the amount of handling.
- Transport recyclable waste with ease and efficiency
Specifications
151.4 L | |
23.5 x 23.5 x 28.75 in. (59.69 x 59.69 x 73 cm) | |
59.69 cm | |
59.69 cm | |
Can serve as central collection containers or shipping containers, reducing the amount of handling | |
23.5 in. |
40 gal. | |
73 cm | |
Square | |
Recycle symbol | |
23.5 in. | |
28.75 in. |