missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium Chloride, Metal min. 99%., MP Biomedicals

Solutions may be slightly hazy

Supplier:  MP Biomedicals Inc 0210282510

 View more versions of this product

Catalog No. ICN10282510


Explore available promotions

Description

Description

Specifications

Specifications

Rubidium Chloride
10 g
ClRb
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
[Cl-].[Rb+]
120.92
CHEBI:78672
7791-11-9
Metal min. 99%.
MFCD00011187
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
rubidium(1+) chloride
62683
120.9

Chemical Identifiers

7791-11-9
120.92
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
62683
rubidium(1+) chloride
ClRb
MFCD00011187
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
CHEBI:78672
[Cl-].[Rb+]
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions