missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium nitrate, 99.8%, pure, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 300070100

 View more versions of this product

Catalog No. AC300070100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Rubidium nitrate
13126-12-0
99.80%
RbNO3
rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate
RTHYXYOJKHGZJT-UHFFFAOYSA-N
rubidium(1+);nitrate
25731
99.8%
Glass bottle
3.1100g/mL
Pure
10g
NO3Rb
MFCD00011193
Solubility in water: 44.28 g/100mL (16°C). Other solubilities: slightly soluble in acetone, very soluble in hno3
[Rb+].[O-][N+]([O-])=O
147.47
147.48
Pure
3.11

Chemical Identifiers

13126-12-0
147.47
RTHYXYOJKHGZJT-UHFFFAOYSA-N
25731
[Rb+].[O-][N+]([O-])=O
NO3Rb
MFCD00011193
rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate
rubidium(1+);nitrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 236-060-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only